Anti-Integrin alpha 3 (P1B5) Mouse Monoclonal Antibody
Lieferant: Oncogene
CP11L-100EA 457 EUR
CP11L-100
Anti-Integrin alpha 3 (P1B5) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Integrin alpha 3 (P1B5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human