Anti-Fibroblast Antigen (AS02) Mouse Monoclonal Antibody

Lieferant: Oncogene
CP28-200EA 504 EUR
CP28-200
Anti-Fibroblast Antigen (AS02) Mouse Monoclonal Antibody
Type: Primary
Antigen: Fibroblast Antigen (AS02)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR