Anti-IgG Peroxidase Conjugate Goat Polyclonal Antibody
Lieferant: Oncogene
DC01L-200EA 172 EUR
DC01L-200
Anti-IgG Peroxidase Conjugate Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IgG Peroxidase Conjugate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat