Anti-Granulocyte Colony Stimulating Factor Mouse Monoclonal Antibody [clone: 5.24]

Lieferant: Oncogene
GF05-200EA 444 EUR
GF05-200
Anti-Granulocyte Colony Stimulating Factor Mouse Monoclonal Antibody [clone: 5.24]
Antikörper
Type: Primary
Antigen: Granulocyte Colony Stimulating Factor (Ab-1) (5.24)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR