Anti-EGFR (Ab-3) (225) Mouse Monoclonal Antibody [clone: 225]

Lieferant: Oncogene
GR13-100EA 572 EUR
GR13-100
Anti-EGFR (Ab-3) (225) Mouse Monoclonal Antibody [clone: 225]
Antikörper
Type: Primary
Antigen: EGFR (Ab-3) (225)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR