Anti-EGFR (Ab-5) (EGFR.1) Mouse Monoclonal Antibody [clone: EGFR.1]

Lieferant: Oncogene
GR15-100EA 444 EUR
GR15-100
Anti-EGFR (Ab-5) (EGFR.1) Mouse Monoclonal Antibody [clone: EGFR.1]
Antikörper
Type: Primary
Antigen: EGFR (Ab-5) (EGFR.1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR