Anti-VDR Rat Monoclonal Antibody [clone: 9A7gammaE10.4]

Lieferant: Oncogene
GR37-100EA 558 EUR
GR37-100
Anti-VDR Rat Monoclonal Antibody [clone: 9A7gammaE10.4]
Antikörper
Type: Primary
Antigen: VDR (vitamin D (1,25- dihydroxyvitamin D3) receptor)
Clonality: monoclonal
Clone: 9A7gammaE10.4
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR