Anti-MTA3 (400-450) Rabbit Polyclonal Antibody

Lieferant: Oncogene
IM1012-50EA 250 EUR
IM1012-50
Anti-MTA3 (400-450) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MTA3 (400-450)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR