Anti-MT1-MMP (Ab-3) (114-6G6) Mouse Monoclonal Antibody

Lieferant: Oncogene
IM42-100EA 432 EUR
IM42-100
Anti-MT1-MMP (Ab-3) (114-6G6) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MT1-MMP (Ab-3) (114-6G6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR