Anti-MMP-2 (Ab-8) (VB3) Mouse Monoclonal Antibody [clone: VB3]

Lieferant: Oncogene
IM68-100EA 444 EUR
IM68-100
Anti-MMP-2 (Ab-8) (VB3) Mouse Monoclonal Antibody [clone: VB3]
Antikörper
Type: Primary
Antigen: MMP-2 (Ab-8) (VB3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR