Anti-MMP-7 (Ab-3) (ID2) Mouse Monoclonal Antibody [clone: ID2]

Lieferant: Oncogene
IM71-100EA 432 EUR
IM71-100
Anti-MMP-7 (Ab-3) (ID2) Mouse Monoclonal Antibody [clone: ID2]
Antikörper
Type: Primary
Antigen: MMP-7 (Ab-3) (ID2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR