Anti-Replication Protein A (Ab-1) (RPA70-9) Mouse Monoclonal Antibody

Lieferant: Oncogene
NA13-100EA 421 EUR
NA13-100
Anti-Replication Protein A (Ab-1) (RPA70-9) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Replication Protein A (Ab-1) (RPA70-9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR