Anti-Pan-Neuronal Neurofilament Marker (SMI-311) Mouse Monoclonal Antibody [clone: SMI-311]

Lieferant: Oncogene
NE1017-100EA 432 EUR
NE1017-100
Anti-Pan-Neuronal Neurofilament Marker (SMI-311) Mouse Monoclonal Antibody [clone: SMI-311]
Antikörper
Type: Primary
Antigen: Pan-Neuronal Neurofilament Marker (SMI-311)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR