Anti-Ryanodine Receptor (C3-33) Mouse Monoclonal Antibody [clone: C3-33]
Lieferant: Oncogene
NR07-100EA 561 EUR
NR07-100
Anti-Ryanodine Receptor (C3-33) Mouse Monoclonal Antibody [clone: C3-33]
Antikörper
Type: Primary
Antigen: Ryanodine Receptor (C3-33)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen