Anti-c-ErbB3 (Ab-5) (H3.105.5) Mouse Monoclonal Antibody [clone: H3.105.5]

Lieferant: Oncogene
OP120-100EA 522 EUR
OP120-100
Anti-c-ErbB3 (Ab-5) (H3.105.5) Mouse Monoclonal Antibody [clone: H3.105.5]
Antikörper
Type: Primary
Antigen: c-ErbB3 (Ab-5) (H3.105.5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR