Anti-IKBKB Mouse Monoclonal Antibody [clone: 10AG2]

Lieferant: Oncogene
OP134-100EA 378 EUR
OP134-100
Anti-IKBKB Mouse Monoclonal Antibody [clone: 10AG2]
Antikörper
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: 10AG2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR