Anti-MDM2 (Ab-4) (2A9C1.18) Mouse Monoclonal Antibody [clone: 2A9C1.18]

Lieferant: Oncogene
OP144-100EA 432 EUR
OP144-100
Anti-MDM2 (Ab-4) (2A9C1.18) Mouse Monoclonal Antibody [clone: 2A9C1.18]
Antikörper
Type: Primary
Antigen: MDM2 (Ab-4) (2A9C1.18)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR