Anti-Pan-Ras Val 12 (Ab-1) (DWP) Mouse Monoclonal Antibody [clone: DWP]

Lieferant: Oncogene
OP38-100EA 432 EUR
OP38-100
Anti-Pan-Ras Val 12 (Ab-1) (DWP) Mouse Monoclonal Antibody [clone: DWP]
Antikörper
Type: Primary
Antigen: Pan-Ras Val 12 (Ab-1) (DWP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR