Anti-Rb (Ab-5) (LM95.1) Mouse Monoclonal Antibody [clone: LM95.1]
Lieferant: Oncogene
OP66-100EA 346 EUR
OP66-100
Anti-Rb (Ab-5) (LM95.1) Mouse Monoclonal Antibody [clone: LM95.1]
Antikörper
Type: Primary
Antigen: Rb (Ab-5) (LM95.1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human