Anti-c-Fos (Ab-2) (4-17) Rabbit Polyclonal Antibody
Lieferant: Oncogene
PC05L-100EA 421 EUR
PC05L-100
Anti-c-Fos (Ab-2) (4-17) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: c-Fos (Ab-2) (4-17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen