Anti-ATM (Ab-3) (819-844) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC116-100EA 421 EUR
PC116-100
Anti-ATM (Ab-3) (819-844) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATM (Ab-3) (819-844)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR