Anti-APP Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC150-25EA 540 EUR
PC150-25
Anti-APP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR