Anti-GABA Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC213L-100EA 476 EUR
PC213L-100
Anti-GABA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR