Anti-FLT1 Goat Polyclonal Antibody

Lieferant: Oncogene
PC322L-100EA 830 EUR
PC322L-100
Anti-FLT1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR