Anti-p53 (Ab-7) (Pantropic) Sheep Polyclonal Antibody

Lieferant: Oncogene
PC35-1EA 620 EUR
PC35-1
Anti-p53 (Ab-7) (Pantropic) Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: p53 (Ab-7) (Pantropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR