Anti-PCNA (247-261) (Ab-5) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC474-50EA 490 EUR
PC474-50
Anti-PCNA (247-261) (Ab-5) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCNA (247-261) (Ab-5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR