Anti-TACE (807 - 823) Rabbit Polyclonal Antibody

PC491-100EA 389 EUR
PC491-100
Anti-TACE (807 - 823) Rabbit Polyclonal Antibody
Type: Primary
Antigen: TACE (807 - 823)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR