Anti-Porin (Ab-5) (185-197) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC548-50EA 378 EUR
PC548-50
Anti-Porin (Ab-5) (185-197) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Porin (Ab-5) (185-197)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR