Anti-MLH1 (Ab-2) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC56-100EA 544 EUR
PC56-100
Anti-MLH1 (Ab-2) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MLH1 (Ab-2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR