Anti-MSH2 (Ab-3) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC57-100EA 457 EUR
PC57-100
Anti-MSH2 (Ab-3) Rabbit Polyclonal Antibody
Type: Primary
Antigen: MSH2 (Ab-3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR