Anti-Caspase-3, Cleaved (Ab-2) Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC679-50EA 421 EUR
PC679-50
Anti-Caspase-3, Cleaved (Ab-2) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Caspase-3, Cleaved (Ab-2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR