Anti-FAS Rabbit Polyclonal Antibody

Lieferant: Oncogene
PC69-100EA 410 EUR
PC69-100
Anti-FAS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR