Anti-Src Family Rabbit Polyclonal Antibody

Lieferant: Oncogene
PK1109-50EA 302 EUR
PK1109-50
Anti-Src Family Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Src Family
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR