Anti-JAK2 Rabbit Polyclonal Antibody
Lieferant: Oncogene
PS1014-100EA 486 EUR
PS1014-100
Anti-JAK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen