Anti-MAPK8/9 Rabbit Polyclonal Antibody

Lieferant: Oncogene
PS1019-100EA 562 EUR
PS1019-100
Anti-MAPK8/9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK8/9 (mitogen-activated protein kinase 8/9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR