Anti-GSK-3 alpha/beta Rabbit Polyclonal Antibody

Lieferant: Oncogene
ST1013-10EA 497 EUR
ST1013-10
Anti-GSK-3 alpha/beta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSK-3 alpha/beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR