Anti-20S Proteasome alpha 1, 2, 3, 5, 6,7-Subunits (MCP231) Mouse Monoclonal Antibody
Lieferant: Oncogene
ST1049-100EA 432 EUR
ST1049-100
Anti-20S Proteasome alpha 1, 2, 3, 5, 6,7-Subunits (MCP231) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: 20S Proteasome alpha 1, 2, 3, 5, 6,7-Subunits (MCP231)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen