Anti-TIAM1 Rabbit Polyclonal Antibody

Lieferant: Oncogene
ST1070-50EA 315 EUR
ST1070-50
Anti-TIAM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIAM1 (T-cell lymphoma invasion and metastasis 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR