Anti-6x-His Tag, Penta-His, Tetra-His and RGS-His Mouse monoclonal antibody unconjugated

733-1738EA 0 EUR
PRME2400300
Anti-6x-His Tag, Penta-His, Tetra-His and RGS-His Mouse monoclonal antibody unconjugated
Type: Primary
Antigen: 6x-His Tag, Penta-His, Tetra-His and RGS-His
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR