Anti-AIMP2 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB11049EA 282 EUR
QEDB11049
Anti-AIMP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR