Anti-KDELR1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11077EA 364 EUR
QEDB11077
Anti-KDELR1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: KDELR1 (KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR