Anti-Protein Disulfide Isomerase (PDI) Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB11081EA 360 EUR
QEDB11081
Anti-Protein Disulfide Isomerase (PDI) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Protein Disulfide Isomerase (PDI)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR