Anti-CRYAB Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB11083EA 282 EUR
QEDB11083
Anti-CRYAB Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR