Anti-TNFRSF10B Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB11092EA 282 EUR
QEDB11092
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR