Anti-CIDEC Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB11093EA 391 EUR
QEDB11093
Anti-CIDEC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CIDEC (cell death-inducing DFFA-like effector c)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR