Anti-HSPA13 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11096EA 330 EUR
QEDB11096
Anti-HSPA13 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR