Anti-HSP90AA1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB11098EA 238 EUR
QEDB11098
Anti-HSP90AA1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSP90AA1 (heat shock protein 90kDa alpha (cytosolic), class A member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR