Anti-HSPD1 Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB11101EA 289 EUR
QEDB11101
Anti-HSPD1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen