Anti-FKBP4 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB11103EA 505 EUR
QEDB11103
Anti-FKBP4 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FKBP4 (FK506 binding protein 4, 59kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR