Anti-HSF2 Rat Monoclonal Antibody
Lieferant: QED Bioscience
QEDB11105EA 418 EUR
QEDB11105
Anti-HSF2 Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSF2 (heat shock transcription factor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestellen